Assay Details
Target Gene Details
Entrez Gene ID: | 35 |
Gene Name: | acyl-CoA dehydrogenase, C-2 to C-3 short chain |
Gene Aliases: |
ACAD3, SCAD |
Location: |
Chr.12:120725738-120740008 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACADS | NM_000017.3 | 10 | 1326 | NP_000008.1 |
| NM_001302554.1 | 10 | 1314 | NP_001289483.1 | |
| AB527081.1 | 10 | 1178 | ||
| AK300645.1 | 10 | 1183 | ||
| BC025963.1 | 10 | 1225 | AAH25963.1 | |
| BU682624.1 | 1 | 639 | ||
| M26393.1 | 10 | 1210 | AAA60307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| esv3630931 | Chr.12:120734778 - 120750697 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map