Assay Details
Target Gene Details
Entrez Gene ID: | 51232 |
Gene Name: | cysteine rich transmembrane BMP regulator 1 |
Gene Aliases: |
CRIM-1, S52 |
Location: |
Chr.2:36356088-36551135 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRIM1 | NM_016441.2 | 1 | 207 | NP_057525.1 |
| XM_005264357.4 | 1 | 203 | XP_005264414.1 | |
| XM_011532898.2 | 1 | 205 | XP_011531200.1 | |
| XM_011532899.2 | 1 | 206 | XP_011531201.1 | |
| XM_011532901.2 | 1 | 206 | XP_011531203.1 | |
| XM_017004258.1 | 1 | 205 | XP_016859747.1 | |
| XM_017004259.1 | 1 | 346 | XP_016859748.1 | |
| XM_017004260.1 | 1 | 203 | XP_016859749.1 | |
| AB209139.1 | 1 | 207 | BAD92376.1 | |
| AF167706.1 | 1 | 180 | AAF34409.1 | |
| AY358371.1 | 1 | 508 | AAQ88737.1 | |
| BC111989.1 | 1 | 178 | AAI11990.1 | |
| BC113371.1 | 1 | 178 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759040 | Chr.2:36016309 - 36363515 on Build GRCh38 | Loss |
|
| nsv2680 | Chr.2:36337775 - 36368588 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map