Assay Details
Target Gene Details
Entrez Gene ID: | 5058 |
Gene Name: | p21 (RAC1) activated kinase 1 |
Gene Aliases: |
PAKalpha |
Location: |
Chr.11:77322015-77474063 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK1 | NM_001128620.1 | 16 | 2510 | NP_001122092.1 |
| NM_002576.4 | 15 | 2461 | NP_002567.3 | |
| XM_011545080.2 | 16 | 2217 | XP_011543382.1 | |
| XM_011545081.2 | 16 | 2286 | XP_011543383.1 | |
| XM_011545082.2 | 17 | 2240 | XP_011543384.1 | |
| XM_011545083.2 | 16 | 2507 | XP_011543385.1 | |
| XM_011545084.2 | 16 | 2077 | XP_011543386.1 | |
| XM_017017846.1 | 15 | 1970 | XP_016873335.1 | |
| XM_017017847.1 | 15 | 2168 | XP_016873336.1 | |
| XM_017017848.1 | 15 | 2237 | XP_016873337.1 | |
| XM_017017849.1 | 15 | 2114 | XP_016873338.1 | |
| XM_017017850.1 | 16 | 2177 | XP_016873339.1 | |
| AK055228.1 | 15 | 1922 | ||
| AK293098.1 | 16 | 2166 | ||
| AK296006.1 | ||||
| BC109299.1 | 15 | 2069 | AAI09300.1 | |
| U24152.1 | AAA65441.1 |
More Information
Set Membership: |
|

Genomic Map