Assay Details
Target Gene Details
Entrez Gene ID: | 518 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9) |
Gene Aliases: |
P3 |
Location: |
Chr.2:175176258-175181762 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5G3 | NM_001002258.4 | 4 | 1396 | NP_001002258.1 |
| NM_001190329.1 | 4 | 1960 | NP_001177258.1 | |
| NM_001689.4 | 5 | 1306 | NP_001680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012709 | Chr.2:175023400 - 175840083 on Build GRCh38 | Gain |
|
| nsv3044 | Chr.2:175164750 - 175210063 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map