Assay Details
Target Gene Details
Entrez Gene ID: | 79817 |
Gene Name: | MOB kinase activator 3B |
Gene Aliases: |
C9orf35, MOB1D, MOBKL2B |
Location: |
Chr.9:27325209-27529852 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3B | NM_024761.4 | 4 | 6013 | NP_079037.3 |
| AA149400.1 | 1 | 319 | ||
| AI692878.1 | 1 | 516 | ||
| CB128567.1 | 1 | 444 | ||
| CN260547.1 | 1 | 68 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466337 | Chr.9:27250870 - 27353462 on Build GRCh38 | Loss |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| nsv1019688 | Chr.9:27234186 - 27356376 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map