Assay Details
Target Gene Details
Entrez Gene ID: | 84720 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class O |
Gene Aliases: |
HPMRS2 |
Location: |
Chr.9:35085493-35096601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGO | NM_001201484.1 | NP_001188413.1 | ||
| NM_032634.3 | NP_116023.2 | |||
| NM_152850.3 | NP_690577.2 | |||
| XM_005251619.3 | XP_005251676.1 | |||
| XM_017015222.1 | XP_016870711.1 | |||
| XM_017015223.1 | XP_016870712.1 | |||
| XM_017015224.1 | XP_016870713.1 | |||
| AK074064.1 | BAB84890.1 | |||
| AK090433.1 | BAC03414.1 | |||
| AL833956.1 | CAD38806.1 | |||
| AY358472.1 | AAQ88836.1 | |||
| BC029271.1 | AAH29271.1 | |||
| BC036916.1 | AAH36916.1 | |||
| BC065282.1 | AAH65282.1 | |||
| BI818120.1 | ||||
| CD387014.1 | ||||
| CR991745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map