Assay Details
Target Gene Details
Entrez Gene ID: | 26289 |
Gene Name: | adenylate kinase 5 |
Gene Aliases: |
AK6 |
Location: |
Chr.1:77281908-77559969 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK5 | NM_012093.3 | 1 | 817 | NP_036225.2 |
| NM_174858.2 | NP_777283.1 | |||
| XM_005270739.4 | XP_005270796.1 | |||
| XM_006710572.3 | XP_006710635.1 | |||
| XM_017001008.1 | XP_016856497.1 | |||
| XM_017001009.1 | 1 | 707 | XP_016856498.1 | |
| XM_017001010.1 | 1 | 1106 | XP_016856499.1 | |
| XM_017001011.1 | XP_016856500.1 | |||
| XM_017001012.1 | XP_016856501.1 | |||
| AF445193.1 | AAP97322.1 | |||
| AK090967.1 | BAC03558.1 | |||
| AY171600.2 | 1 | 822 | AAO16520.2 | |
| BC033896.1 | 1 | 822 | AAH33896.1 | |
| BC036666.1 | AAH36666.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79754897] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map