Assay Details
Target Gene Details
Entrez Gene ID: | 55212 |
Gene Name: | Bardet-Biedl syndrome 7 |
Gene Aliases: |
BBS2L1 |
Location: |
Chr.4:121824329-121870520 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS7 | NM_018190.3 | NP_060660.2 | ||
| NM_176824.2 | NP_789794.1 | |||
| XM_005263106.3 | XP_005263163.1 | |||
| XM_011532079.2 | XP_011530381.1 | |||
| XM_011532080.2 | XP_011530382.1 | |||
| XM_011532081.2 | XP_011530383.1 | |||
| XM_017008357.1 | XP_016863846.1 | |||
| XM_017008358.1 | XP_016863847.1 | |||
| AF521643.1 | AAO16025.1 | |||
| AF521644.1 | AAO16026.1 | |||
| AK001577.1 | BAA91767.1 | |||
| AK308076.1 | ||||
| BC032691.1 | AAH32691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map