Assay Details
Target Gene Details
Entrez Gene ID: | 347734 |
Gene Name: | solute carrier family 35 member B2 |
Gene Aliases: |
PAPST1, SLL, UGTrel4 |
Location: |
Chr.6:44254101-44257890 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35B2 | NM_001286509.1 | 4 | 1543 | NP_001273438.1 |
| NM_001286510.1 | 4 | 1542 | NP_001273439.1 | |
| NM_001286511.1 | 4 | 1497 | NP_001273440.1 | |
| NM_001286512.1 | 4 | 1546 | NP_001273441.1 | |
| NM_001286513.1 | 3 | 1392 | NP_001273442.1 | |
| NM_001286517.1 | 3 | 1353 | NP_001273446.1 | |
| NM_001286519.1 | 2 | 1198 | NP_001273448.1 | |
| NM_001286520.1 | 4 | 1551 | NP_001273449.1 | |
| NM_178148.3 | 4 | 1547 | NP_835361.1 | |
| AB097021.1 | 5 | 1384 | BAC77374.1 | |
| AB097039.1 | 5 | 1384 | BAC77392.1 | |
| AK075456.1 | 4 | 1476 | BAC11631.1 | |
| AK123749.1 | 4 | 1444 | ||
| AK223028.1 | 3 | 1197 | BAD96748.1 | |
| AK293251.1 | 2 | 1165 | ||
| AK293344.1 | 3 | 1349 | ||
| AK307572.1 | ||||
| AY491520.1 | 4 | 1513 | AAS79661.1 | |
| BC016839.1 | 1 | 570 | AAH16839.1 | |
| BC024288.1 | 4 | 1511 | AAH24288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524398 | Chr.6:44233216 - 44286028 on Build GRCh38 | Loss |
|
| nsv5282 | Chr.6:44217138 - 44259154 on Build GRCh38 | Insertion |
|
| nsv603003 | Chr.6:44224183 - 44286028 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map