Assay Details
Target Gene Details
Entrez Gene ID: | 79828 |
Gene Name: | methyltransferase like 8 |
Gene Aliases: |
TIP |
Location: |
Chr.2:171315746-171434803 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL8 | NM_001321154.1 | 10 | 1885 | NP_001308083.1 |
| NM_001321155.1 | 11 | 2009 | NP_001308084.1 | |
| NM_001321156.1 | 11 | 2079 | NP_001308085.1 | |
| NM_001321157.1 | 11 | 1912 | NP_001308086.1 | |
| NM_001321158.1 | 9 | 1633 | NP_001308087.1 | |
| NM_001321159.1 | 9 | 1730 | NP_001308088.1 | |
| NM_001321160.1 | 10 | 1902 | NP_001308089.1 | |
| NM_001321161.1 | 8 | 1526 | NP_001308090.1 | |
| NM_001321162.1 | 9 | 1747 | NP_001308091.1 | |
| NM_024770.4 | 10 | 1788 | NP_079046.2 | |
| NR_135568.1 | 7 | 1130 | ||
| XM_011511875.2 | 10 | 1700 | XP_011510177.1 | |
| XM_017004984.1 | 11 | 1928 | XP_016860473.1 | |
| XM_017004985.1 | 11 | 2026 | XP_016860474.1 | |
| XM_017004986.1 | 10 | 1804 | XP_016860475.1 | |
| XM_017004987.1 | 9 | 1666 | XP_016860476.1 | |
| XM_017004988.1 | 9 | 1650 | XP_016860477.1 | |
| XM_017004989.1 | 10 | 1819 | XP_016860478.1 | |
| AK024046.1 | 9 | 1748 | BAB14797.1 | |
| AK093218.1 | ||||
| AK124092.1 |
More Information
Set Membership: |
|

Genomic Map