Assay Details
Target Gene Details
Entrez Gene ID: | 27299 |
Gene Name: | ADAM like decysin 1 |
Gene Aliases: |
M12.219 |
Location: |
Chr.8:24384285-24406013 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMDEC1 | NM_001145271.1 | NP_001138743.1 | ||
| NM_001145272.1 | NP_001138744.1 | |||
| NM_014479.3 | NP_055294.1 | |||
| AK300959.1 | ||||
| AK301007.1 | ||||
| AK311528.1 | ||||
| AK316320.1 | ||||
| AK316322.1 | ||||
| BC069582.1 | AAH69582.1 | |||
| BC074877.2 | AAH74877.1 | |||
| BC074878.2 | AAH74878.1 | |||
| DC387711.1 | ||||
| Y13323.2 | CAA73764.2 |
Target Gene Details
Entrez Gene ID: | 101929294 |
Gene Name: | uncharacterized LOC101929294 |
Gene Aliases: |
- |
Location: |
Chr.8:24295814-24548618 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929294 | NR_125808.1 | |||
| DA221074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891377 | Chr.8:23722497 - 25043041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map