Assay Details
Target Gene Details
Entrez Gene ID: | 23259 |
Gene Name: | DDHD domain containing 2 |
Gene Aliases: |
SAMWD1, SPG54, iPLA(1)gamma |
Location: |
Chr.8:38231343-38273442 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDHD2 | XM_011544456.1 | XP_011542758.1 |
Target Gene Details
Entrez Gene ID: | 84513 |
Gene Name: | phospholipid phosphatase 5 |
Gene Aliases: |
DPPL1, HTPAP, PPAPDC1B |
Location: |
Chr.8:38263130-38269435 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP5 | NM_001102559.1 | NP_001096029.1 | ||
| NM_032483.3 | NP_115872.2 | |||
| XM_005273661.4 | XP_005273718.2 | |||
| XM_011544671.1 | XP_011542973.1 | |||
| XM_011544672.1 | XP_011542974.1 | |||
| XM_017013907.1 | XP_016869396.1 | |||
| AF212238.1 | AAK14924.1 | |||
| AK292118.1 | ||||
| BC033025.1 | AAH33025.1 | |||
| BC106014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7164n100 | Chr.8:38194164 - 38310837 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map