Assay Details
Target Gene Details
Entrez Gene ID: | 7024 |
Gene Name: | transcription factor CP2 |
Gene Aliases: |
LBP1C, LSF, LSF1D, SEF, TFCP2C |
Location: |
Chr.12:51093756-51173143 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFCP2 | NM_001173452.1 | NP_001166923.1 | ||
| NM_001173453.1 | NP_001166924.1 | |||
| NM_005653.4 | NP_005644.2 | |||
| AK291264.1 | ||||
| AK308087.1 | ||||
| BC003634.2 | AAH03634.1 | |||
| M84810.1 | ||||
| U03494.1 | AAA21324.1 | |||
| U03495.1 | AAA21325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115299696] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map