Assay Details
Target Gene Details
Entrez Gene ID: | 51111 |
Gene Name: | lysine methyltransferase 5B |
Gene Aliases: |
CGI-85, CGI85, SUV420H1 |
Location: |
Chr.11:68154863-68213904 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT5B | NM_001300907.1 | 12 | 3834 | NP_001287836.1 |
| NM_001300908.1 | 11 | 3801 | NP_001287837.1 | |
| NM_017635.4 | 11 | 3766 | NP_060105.3 | |
| XM_005274035.3 | 11 | 3986 | XP_005274092.2 | |
| XM_005274036.3 | 10 | 3917 | XP_005274093.2 | |
| XM_006718581.1 | 10 | 3648 | XP_006718644.1 | |
| XM_011545091.1 | 10 | 3560 | XP_011543393.1 | |
| XM_011545092.2 | 10 | 3750 | XP_011543394.1 | |
| XM_011545093.2 | 11 | 4031 | XP_011543395.1 | |
| XM_017017876.1 | 12 | 4347 | XP_016873365.1 | |
| XM_017017877.1 | 12 | 4133 | XP_016873366.1 | |
| XM_017017878.1 | 12 | 4078 | XP_016873367.1 | |
| XM_017017879.1 | 12 | 3879 | XP_016873368.1 | |
| AI659791.1 | 1 | 150 | ||
| BG697990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825966 | Chr.11:68156334 - 68157035 on Build GRCh38 | Gain |
|
| nsv825963 | Chr.11:67996205 - 68429586 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map