Assay Details
Target Gene Details
Entrez Gene ID: | 92483 |
Gene Name: | lactate dehydrogenase A like 6B |
Gene Aliases: |
LDH6B, LDHAL6, LDHL |
Location: |
Chr.15:59206816-59208588 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDHAL6B | NM_033195.2 | 1 | 52 | NP_149972.1 |
| AK058192.1 | BAB71710.1 | |||
| AK312777.1 | ||||
| AY009108.1 | AAG49399.1 | |||
| AY642121.1 | 1 | 45 | AAT65080.1 | |
| BC022034.1 | AAH22034.1 |
Target Gene Details
Entrez Gene ID: | 4643 |
Gene Name: | myosin IE |
Gene Aliases: |
FSGS6, HuncM-IC, MYO1C |
Location: |
Chr.15:59135969-59372872 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO1E | NM_004998.3 | NP_004989.2 | ||
| AB290177.1 | ||||
| AK291688.1 | ||||
| BC098392.1 | AAH98392.1 | |||
| U14391.1 | AAA62667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| nsv833027 | Chr.15:59182607 - 59316345 on Build GRCh38 | Gain+Loss |
|
| nsv974584 | Chr.15:59206826 - 59208117 on Build GRCh38 | Duplication |
|
| nsv833025 | Chr.15:59043949 - 59229977 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map