Assay Details
Target Gene Details
Entrez Gene ID: | 55233 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
Location: |
Chr.2:74152598-74178967 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB1A | NM_001317110.1 | 6 | 2101 | NP_001304039.1 |
| NM_001317111.1 | 6 | 1997 | NP_001304040.1 | |
| NM_001317112.1 | 5 | 1940 | NP_001304041.1 | |
| NM_018221.4 | 6 | 2104 | NP_060691.2 | |
| XM_011532948.2 | 5 | 1800 | XP_011531250.1 | |
| AK001650.1 | 6 | 1994 | BAA91810.1 |
More Information
Set Membership: |
|

Genomic Map