Assay Details
Target Gene Details
Entrez Gene ID: | 351 |
Gene Name: | amyloid beta precursor protein |
Gene Aliases: |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2 |
Location: |
Chr.21:25880550-26171128 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APP | NM_000484.3 | 18 | 2600 | NP_000475.1 |
NM_001136016.3 | 17 | 2524 | NP_001129488.1 | |
NM_001136129.2 | 15 | 2207 | NP_001129601.1 | |
NM_001136130.2 | 17 | 2432 | NP_001129602.1 | |
NM_001136131.2 | 16 | 2262 | NP_001129603.1 | |
NM_001204301.1 | 17 | 2546 | NP_001191230.1 | |
NM_001204302.1 | 16 | 2489 | NP_001191231.1 | |
NM_001204303.1 | 15 | 2321 | NP_001191232.1 | |
NM_201413.2 | 17 | 2543 | NP_958816.1 | |
NM_201414.2 | 16 | 2375 | NP_958817.1 | |
A02759.1 | 16 | 2321 | CAA00268.1 | |
AF282245.1 | 15 | 2134 | AAQ14327.1 | |
AK294534.1 | 17 | 2379 | ||
AK295373.1 | 8 | 1100 | ||
AK295621.1 | 17 | 2485 | ||
AK296229.1 | 16 | 2262 | ||
AK297229.1 | 14 | 1738 | ||
AK297412.1 | 15 | 2104 | ||
AK298861.1 | 15 | 2277 | ||
BC065529.1 | 17 | 2474 | AAH65529.1 | |
M15533.1 | 3 | 381 | AAA35540.1 | |
M16765.1 | 11 | 1335 | AAA51722.1 | |
M18734.1 | 7 | 883 | AAA51726.1 | |
X06989.1 | 17 | 2467 | CAA30050.1 | |
Y00264.1 | 16 | 2322 | CAA68374.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
