Assay Details
Target Gene Details
Entrez Gene ID: | 54918 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 6 |
Gene Aliases: |
CKLFSF6, PRO2219 |
Location: |
Chr.3:32481312-32502911 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM6 | NM_017801.2 | 4 | 2854 | NP_060271.1 |
| AF116694.1 | 1 | 564 | AAF71114.1 | |
| AF479261.1 | 4 | 3351 | AAN73039.1 | |
| AK172793.1 | 1 | 1732 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
| nsv834652 | Chr.3:32375792 - 32564922 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map