Assay Details
Target Gene Details
Entrez Gene ID: | 50839 |
Gene Name: | taste 2 receptor member 10 |
Gene Aliases: |
T2R10, TRB2 |
Location: |
Chr.12:10825346-10826269 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS2R10 | NM_023921.1 | 1 | 65 | NP_076410.1 |
| BC069089.1 | 1 | 65 | AAH69089.1 | |
| BC101762.1 | 1 | 154 | AAI01763.1 | |
| BC101764.1 | 1 | 154 | AAI01765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628543 | Chr.12:10720386 - 10881212 on Build GRCh38 | Gain |
|
| esv2759879 | Chr.12:10817021 - 11561987 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map