Assay Details
Target Gene Details
Entrez Gene ID: | 50808 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AK3L1, AK6, AKL3L, AKL3L1, FIX |
Location: |
Chr.9:4709556-4742043 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK3 | NM_001199852.1 | NP_001186781.1 | ||
| NM_001199853.1 | NP_001186782.1 | |||
| NM_001199855.1 | NP_001186784.1 | |||
| NM_001199856.1 | NP_001186785.1 | |||
| NM_016282.3 | NP_057366.2 | |||
| AB021870.1 | BAA87913.1 | |||
| AF086913.1 | AAP97143.1 | |||
| AF100785.1 | AAP97224.1 | |||
| AF183419.1 | AAG09688.1 | |||
| AK001553.1 | BAA91753.1 | |||
| AK001951.1 | BAA91996.1 | |||
| AK027534.1 | BAB55183.1 | |||
| AK098205.1 | ||||
| AK124824.1 | ||||
| AK298200.1 | ||||
| BC013771.1 | AAH13771.1 | |||
| BI550730.1 | ||||
| BI601245.1 | ||||
| DA570364.1 | ||||
| DA718987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015376 | Chr.9:4686223 - 4737890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map