Assay Details
Target Gene Details
| Entrez Gene ID: | 4807 | 
| Gene Name: | nescient helix-loop-helix 1 | 
| Gene Aliases: | HEN1, NSCL, NSCL1, bHLHa35 | 
| Location: | Chr.1:160367071-160372848 on Build GRCh38 | 
| Assay Gene Location: | Overlaps - Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NHLH1 | NM_005598.3 | NP_005589.1 | ||
| BC013789.1 | AAH13789.1 | |||
| M96739.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv548056 | Chr.1:160363952 - 160422364 on Build GRCh38 | Loss |  NHLH1  VANGL2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top