Assay Details
Target Gene Details
Entrez Gene ID: | 5000 |
Gene Name: | origin recognition complex subunit 4 |
Gene Aliases: |
ORC4L, ORC4P |
Location: |
Chr.2:147930397-148021604 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORC4 | NM_001190879.2 | 15 | 2918 | NP_001177808.1 |
| NM_001190881.2 | 11 | 2536 | NP_001177810.1 | |
| NM_001190882.2 | 13 | 2680 | NP_001177811.1 | |
| NM_002552.4 | 14 | 2802 | NP_002543.2 | |
| NM_181741.3 | 14 | 2911 | NP_859525.1 | |
| NM_181742.3 | 14 | 2806 | NP_859526.1 | |
| AK128860.1 | 1 | 133 | ||
| BM468640.1 | 1 | 580 | ||
| BQ716497.1 | ||||
| DA120022.1 | 1 | 174 |
More Information
Set Membership: |
|

Genomic Map