Assay Details
Target Gene Details
Entrez Gene ID: | 116496 |
Gene Name: | family with sequence similarity 129 member A |
Gene Aliases: |
C1orf24, GIG39, NIBAN |
Location: |
Chr.1:184791025-184975132 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM129A | NM_052966.3 | 14 | 4375 | NP_443198.1 |
| XM_011509140.2 | 15 | 5250 | XP_011507442.1 | |
| XM_011509141.2 | 14 | 5139 | XP_011507443.1 | |
| AF288391.1 | 14 | 4339 | AAG60611.1 | |
| AK074139.1 | 2 | 2583 | BAB84965.1 | |
| AL137572.1 | 1 | 999 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832048 | Chr.1:184691012 - 184889668 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map