Assay Details
Target Gene Details
Entrez Gene ID: | 145264 |
Gene Name: | serpin family A member 12 |
Gene Aliases: |
OL-64 |
Location: |
Chr.14:94481651-94517844 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINA12 | NM_001304461.1 | 2 | 711 | NP_001291390.1 |
| NM_173850.3 | 3 | 1284 | NP_776249.1 | |
| XM_011536451.2 | 2 | 687 | XP_011534753.1 | |
| XM_011536452.2 | 2 | 681 | XP_011534754.1 | |
| XM_011536453.2 | 2 | 593 | XP_011534755.1 | |
| XM_011536454.2 | 2 | 672 | XP_011534756.1 | |
| XM_011536455.1 | 1 | 505 | XP_011534757.1 | |
| XM_017020989.1 | 2 | 682 | XP_016876478.1 | |
| XM_017020990.1 | 2 | 667 | XP_016876479.1 | |
| AY177692.1 | 1 | 506 | AAO18649.1 | |
| AY326420.1 | 2 | 711 | AAP88384.1 | |
| BC040857.1 | 3 | 1284 | AAH40857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832863 | Chr.14:94348482 - 94501487 on Build GRCh38 | Gain |
|
| nsv510649 | Chr.14:94421103 - 94544514 on Build GRCh38 | Deletion |
|
| nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
|
| nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
|
| nsv565624 | Chr.14:94494633 - 94512931 on Build GRCh38 | Loss |
|
| nsv832864 | Chr.14:94362101 - 94538150 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map