Assay Details
Target Gene Details
Entrez Gene ID: | 6546 |
Gene Name: | solute carrier family 8 member A1 |
Gene Aliases: |
NCX1 |
Location: |
Chr.2:40112146-40512452 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Target Gene Details
Entrez Gene ID: | 100128590 |
Gene Name: | SLC8A1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:39917634-40255209 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8A1-AS1 | NR_038441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833892 | Chr.2:39961645 - 40136340 on Build GRCh38 | Loss |
|
| nsv581502 | Chr.2:40046763 - 40232505 on Build GRCh38 | Gain |
|
| esv3575236 | Chr.2:39983042 - 40232432 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map