Assay Details
Target Gene Details
Entrez Gene ID: | 129642 |
Gene Name: | membrane bound O-acyltransferase domain containing 2 |
Gene Aliases: |
LPAAT, LPCAT4, LPEAT, LPLAT 2, OACT2 |
Location: |
Chr.2:8852690-9003750 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBOAT2 | NM_001321265.1 | 12 | 2201 | NP_001308194.1 |
| NM_001321266.1 | 11 | 2123 | NP_001308195.1 | |
| NM_001321267.1 | 14 | 2424 | NP_001308196.1 | |
| NM_138799.3 | 13 | 2347 | NP_620154.2 | |
| NR_135598.1 | 14 | 2399 | ||
| NR_135599.1 | 13 | 2666 | ||
| NR_135600.1 | 13 | 2253 | ||
| NR_135601.1 | 11 | 2042 | ||
| NR_135602.1 | 12 | 2214 | ||
| NR_135603.1 | 14 | 2440 | ||
| NR_135604.1 | 13 | 2363 | ||
| NR_135605.1 | 12 | 2175 | ||
| NR_135606.1 | 13 | 2266 | ||
| AK074779.1 | 3 | 1167 | BAC11204.1 | |
| BC146871.1 | 13 | 2291 | ||
| BC157827.1 | 13 | 2291 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833359 | Chr.2:8810194 - 8990672 on Build GRCh38 | Loss |
|
| nsv517367 | Chr.2:8853375 - 9046387 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116554173] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map