Assay Details
Target Gene Details
Entrez Gene ID: | 55214 |
Gene Name: | prolyl 3-hydroxylase 2 |
Gene Aliases: |
LEPREL1, MCVD, MLAT4 |
Location: |
Chr.3:189956728-190122437 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3H2 | NM_001134418.1 | 15 | 2431 | NP_001127890.1 |
| NM_018192.3 | 15 | 3064 | NP_060662.2 | |
| XM_011512955.1 | 15 | 2338 | XP_011511257.1 | |
| AJ430351.2 | 15 | 2874 | CAD23039.2 | |
| AK001580.1 | 15 | 2265 | BAA91769.1 | |
| AK056447.1 | 15 | 2874 | ||
| AK125134.1 | 15 | 2869 | ||
| BC005029.1 | 15 | 2259 | AAH05029.1 | |
| BU621129.1 | 1 | 549 |
More Information
Set Membership: |
|

Genomic Map