Assay Details
Target Gene Details
Entrez Gene ID: | 1081 |
Gene Name: | glycoprotein hormones, alpha polypeptide |
Gene Aliases: |
CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA |
Location: |
Chr.6:87085498-87095147 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CGA | NM_000735.3 | 3 | 309 | NP_000726.1 |
| NM_001252383.1 | 4 | 402 | NP_001239312.1 | |
| AK311972.1 | 3 | 266 | ||
| BC010957.1 | 3 | 272 | AAH10957.1 | |
| BC020782.1 | 3 | 268 | AAH20782.1 | |
| BC055080.1 | 3 | 270 | AAH55080.1 | |
| BG571584.1 | 4 | 359 | ||
| BG619154.1 | 3 | 310 | ||
| CR457427.1 | 2 | 167 | CAG33708.1 | |
| FJ624078.1 | 2 | 95 | ||
| FJ624079.1 | 2 | 167 | ||
| V00518.1 | 3 | 217 | CAA23777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604106 | Chr.6:85995537 - 87138584 on Build GRCh38 | Gain |
|
| nsv5385 | Chr.6:87071297 - 87093945 on Build GRCh38 | Deletion |
|
| esv1002835 | Chr.6:87082506 - 87090909 on Build GRCh38 | Deletion |
|
| esv2763585 | Chr.6:85938269 - 87211344 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map