Assay Details
Target Gene Details
Entrez Gene ID: | 10314 |
Gene Name: | LanC like 1 |
Gene Aliases: |
GPR69A, p40 |
Location: |
Chr.2:210431249-210477618 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LANCL1 | NM_001136574.1 | 10 | 3786 | NP_001130046.1 |
| NM_001136575.1 | 10 | 3781 | NP_001130047.1 | |
| NM_006055.2 | 10 | 3875 | NP_006046.1 | |
| XM_005246243.2 | 10 | 3896 | XP_005246300.1 | |
| Y11395.1 | 10 | 3815 | CAA72205.1 |
Target Gene Details
Entrez Gene ID: | 102724820 |
Gene Name: | LANCL1 antisense RNA 1 |
Gene Aliases: |
LANCLOT |
Location: |
Chr.2:210324712-210470333 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LANCL1-AS1 | NR_110604.1 | |||
| NR_110605.1 | ||||
| NR_110606.1 | ||||
| AL599033.1 | ||||
| BX119066.1 | ||||
| DA820426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012567 | Chr.2:210222864 - 210450830 on Build GRCh38 | Gain |
|
| nsv3147 | Chr.2:210416755 - 210462120 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map