Assay Details
Target Gene Details
Entrez Gene ID: | 81607 |
Gene Name: | nectin cell adhesion molecule 4 |
Gene Aliases: |
EDSS1, LNIR, PRR4, PVRL4, nectin-4 |
Location: |
Chr.1:161070991-161089595 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECTIN4 | NM_030916.2 | 9 | 2500 | NP_112178.2 |
| XM_005245508.3 | 8 | 2396 | XP_005245565.1 | |
| XM_011510021.2 | 9 | 2474 | XP_011508323.1 | |
| XM_011510022.2 | 8 | 2399 | XP_011508324.1 | |
| AB105191.1 | 9 | 2457 | ||
| AF160477.1 | 9 | 2336 | AAF82399.1 | |
| AF218028.1 | 1 | 674 | AAG17270.1 | |
| AK027753.1 | 9 | 2457 | BAB55344.1 | |
| BC010423.1 | 9 | 2448 | AAH10423.1 | |
| BX641083.1 | 1 | 1288 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3221 | Chr.1:161038385 - 161083189 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80118484] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map