Assay Details
Target Gene Details
Entrez Gene ID: | 11102 |
Gene Name: | ribonuclease P/MRP subunit p14 |
Gene Aliases: |
HsHTD2, P14 |
Location: |
Chr.3:58306245-58322642 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPP14 | NM_001098783.2 | 2 | 445 | NP_001092253.1 |
| NM_007042.4 | 2 | 211 | NP_008973.1 | |
| NR_049755.1 | ||||
| NR_049756.1 | ||||
| NR_049757.1 | ||||
| NR_049758.1 | ||||
| XM_011533313.2 | 2 | 245 | XP_011531615.1 | |
| XM_017005641.1 | 2 | 208 | XP_016861130.1 | |
| AF001175.1 | 2 | 186 | AAD00892.1 | |
| AK311396.1 | ||||
| AK311969.1 | 2 | 140 | ||
| AV760694.2 | ||||
| BC002441.1 | 2 | 159 | AAH02441.1 | |
| BC007342.1 | 2 | 159 | AAH07342.1 | |
| BC012017.1 | 2 | 172 | AAH12017.1 | |
| BP352444.1 | ||||
| BP352480.1 | 2 | 428 | ||
| CD698741.1 | ||||
| CN354601.1 | 2 | 138 | ||
| CR407611.1 | CAG28539.1 | |||
| DA040251.1 | 2 | 175 | ||
| DC303206.1 | ||||
| DC360931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568768 | Chr.3:58306185 - 58316063 on Build GRCh38 | Loss |
|
| nsv528147 | Chr.3:57929747 - 58384450 on Build GRCh38 | Gain |
|
| nsv590369 | Chr.3:58283985 - 58331965 on Build GRCh38 | Loss |
|
| nsv1001814 | Chr.3:58158881 - 58354197 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map