Assay Details
Target Gene Details
Entrez Gene ID: | 83259 |
Gene Name: | protocadherin 11 Y-linked |
Gene Aliases: |
PCDH-PC, PCDH22, PCDHX, PCDHY |
Location: |
Chr.Y:5000044-5742228 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH11Y | NM_001278619.1 | 5 | 777 | NP_001265548.1 |
| NM_032971.2 | 4 | 608 | NP_116753.1 | |
| NM_032972.2 | 1 | 862 | NP_116754.1 | |
| NM_032973.2 | 1 | 862 | NP_116755.1 | |
| XM_017030079.1 | 7 | 5139 | XP_016885568.1 | |
| XM_017030080.1 | 6 | 1091 | XP_016885569.1 | |
| XM_017030081.1 | 5 | 861 | XP_016885570.1 | |
| XM_017030082.1 | 5 | 674 | XP_016885571.1 | |
| XM_017030083.1 | 2 | 438 | XP_016885572.1 | |
| AF277053.1 | 5 | 708 | AAL55729.1 | |
| AF332216.1 | 1 | 862 | AAK13468.1 | |
| AF332217.1 | 1 | 862 | AAK13469.1 | |
| AJ276803.1 | 4 | 608 | CAC13122.1 | |
| AJ564958.1 | 1 | 63 | CAD92429.1 | |
| AK289676.1 | 4 | 399 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577576 | Chr.Y:5026633 - 5785419 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map