Assay Details
Target Gene Details
Entrez Gene ID: | 1601 |
Gene Name: | DAB2, clathrin adaptor protein |
Gene Aliases: |
DOC-2, DOC2 |
Location: |
Chr.5:39371674-39425233 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAB2 | NM_001244871.1 | 14 | 2884 | NP_001231800.1 |
| NM_001343.3 | 15 | 2947 | NP_001334.2 | |
| AF188298.1 | 13 | 2441 | AAF05540.1 | |
| AK092836.1 | 1 | 887 | ||
| AK130136.1 | 5 | 957 | ||
| AK297984.1 | ||||
| BC003064.2 | 15 | 2543 | AAH03064.1 | |
| BU742719.1 | ||||
| U39050.1 | 14 | 2515 | AAC50824.1 | |
| U53446.1 | 15 | 2560 | AAA98975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597860 | Chr.5:38995581 - 40274356 on Build GRCh38 | Gain |
|
| esv3604783 | Chr.5:39235417 - 39382661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map