Assay Details
Target Gene Details
Entrez Gene ID: | 23368 |
Gene Name: | protein phosphatase 1 regulatory subunit 13B |
Gene Aliases: |
ASPP1, p53BP2-like, p85 |
Location: |
Chr.14:103733751-103848967 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R13B | NM_015316.2 | 17 | 3621 | NP_056131.2 |
| XM_005267487.4 | 17 | 4025 | XP_005267544.1 | |
| XM_011536593.2 | 18 | 4130 | XP_011534895.1 | |
| XM_017021116.1 | 18 | 3767 | XP_016876605.1 | |
| XM_017021117.1 | 18 | 3543 | XP_016876606.1 | |
| XM_017021118.1 | 18 | 3456 | XP_016876607.1 | |
| XM_017021119.1 | 12 | 2750 | XP_016876608.1 | |
| AB018314.1 | 13 | 2912 | BAA34491.1 | |
| AJ318887.2 | 17 | 3496 | CAC83011.2 | |
| AK021580.1 | 3 | 396 | ||
| AK023188.1 | 8 | 2137 | ||
| AK125374.1 | 2 | 1800 | ||
| AK125913.1 | 19 | 3992 | BAC86342.1 | |
| AK307149.1 | ||||
| BC136527.1 | 17 | 3425 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470673 | Chr.14:103695926 - 103753612 on Build GRCh38 | Loss |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
| esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map