Assay Details
Target Gene Details
Entrez Gene ID: | 100526694 |
Gene Name: | MSANTD3-TMEFF1 readthrough |
Gene Aliases: |
C9orf3-TMEFF1, C9orf30-TMEFF1 |
Location: |
Chr.9:100441906-100577636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSANTD3-TMEFF1 | NM_001198812.1 | NP_001185741.1 | ||
| AL831919.1 | CAD38580.2 |
Target Gene Details
Entrez Gene ID: | 8577 |
Gene Name: | transmembrane protein with EGF like and two follistatin like domains 1 |
Gene Aliases: |
C9orf2, CT120.1, H7365, TR-1 |
Location: |
Chr.9:100473238-100577636 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEFF1 | NM_003692.4 | 1 | 395 | NP_003683.2 |
| BC035056.1 | 1 | 197 | AAH35056.1 | |
| DN602026.1 | 1 | 395 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825012 | Chr.9:100389923 - 100493999 on Build GRCh38 | Gain |
|
| nsv614955 | Chr.9:100419314 - 100498945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map