Assay Details
Target Gene Details
Entrez Gene ID: | 25800 |
Gene Name: | solute carrier family 39 member 6 |
Gene Aliases: |
LIV-1, ZIP6 |
Location: |
Chr.18:36108531-36129814 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A6 | NM_001099406.1 | 4 | 689 | NP_001092876.1 |
| NM_012319.3 | 5 | 1487 | NP_036451.3 | |
| XM_011525900.1 | 5 | 1920 | XP_011524202.1 | |
| XM_011525901.1 | 6 | 2604 | XP_011524203.1 | |
| AK299102.1 | 4 | 520 | ||
| AK301539.1 | 4 | 639 | ||
| AK304079.1 | 5 | 1411 | ||
| BC008317.2 | 1 | 56 | AAH08317.2 | |
| BC039498.1 | 4 | 680 | AAH39498.1 | |
| BX336036.2 | 4 | 458 | ||
| U41060.2 | 6 | 1316 | AAA96258.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map