Assay Details
Target Gene Details
Entrez Gene ID: | 4100 |
Gene Name: | MAGE family member A1 |
Gene Aliases: |
CT1.1, MAGE1 |
Location: |
Chr.X:153179284-153183878 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA1 | NM_004988.4 | 3 | 271 | NP_004979.3 |
| AF463515.1 | 2 | 191 | AAL69948.1 | |
| AK314979.1 | 3 | 271 | ||
| AY148486.1 | 3 | 229 | AAN62752.1 | |
| BC017555.1 | 3 | 238 | AAH17555.1 | |
| BP314195.1 | 2 | 192 | ||
| BT009789.1 | 1 | 51 | AAP88791.1 | |
| DB053667.1 | 3 | 285 | ||
| EU161102.1 | 3 | 229 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| dgv2n30 | Chr.X:153142213 - 153228848 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map