Assay Details
Target Gene Details
Entrez Gene ID: | 55215 |
Gene Name: | Fanconi anemia complementation group I |
Gene Aliases: |
KIAA1794 |
Location: |
Chr.15:89243954-89317132 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCI | NM_001113378.1 | NP_001106849.1 | ||
| NM_018193.2 | NP_060663.2 | |||
| XM_011521756.2 | XP_011520058.1 | |||
| XM_011521757.2 | XP_011520059.1 | |||
| XM_011521764.2 | XP_011520066.1 | |||
| XM_011521767.2 | XP_011520069.1 | |||
| AK001581.1 | BAA91770.1 | |||
| AK055176.1 | ||||
| BC004277.1 | AAH04277.1 | |||
| BC140769.1 | ||||
| BC144483.1 | ||||
| BM461935.1 | ||||
| BU187299.1 | ||||
| EF469766.1 | ||||
| EF567077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map