Assay Details
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU2 | NR_002612.1 | |||
| AY455757.1 |
Target Gene Details
Entrez Gene ID: | 283518 |
Gene Name: | potassium channel regulator |
Gene Aliases: |
DLTET |
Location: |
Chr.13:50015254-50020922 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNRG | NM_173605.1 | 2 | 1172 | NP_775876.1 |
| NM_199464.2 | 3 | 1270 | NP_955751.1 | |
| AF086244.1 | 1 | 138 | ||
| AY129653.1 | 3 | 1080 | AAN06090.1 | |
| AY129654.1 | 2 | 990 | AAN06091.1 | |
| AY169387.1 | 3 | 1270 | AAO11777.1 | |
| AY169388.1 | 2 | 1172 | AAO11778.1 | |
| BC020887.1 | 3 | 1080 | ||
| BC032343.1 | 2 | 966 | AAH32343.1 | |
| BX116021.1 | 1 | 133 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580637 | Chr.13:50006792 - 50829882 on Build GRCh38 | Loss |
|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map