Assay Details
Target Gene Details
Entrez Gene ID: | 83693 |
Gene Name: | hydroxysteroid dehydrogenase like 1 |
Gene Aliases: |
SDR12C3 |
Location: |
Chr.16:84122139-84145195 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSDL1 | NM_001146051.1 | 7 | 2832 | NP_001139523.1 |
| NM_031463.4 | 6 | 2997 | NP_113651.4 | |
| XM_005256189.2 | 6 | 2965 | XP_005256246.1 | |
| AK025626.1 | 2 | 2108 | ||
| AK074878.1 | 6 | 2961 | BAC11262.1 | |
| AK098199.1 | 1 | 2395 | ||
| AY028377.1 | 5 | 2861 | AAK16927.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain |
|
| nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map