Assay Details
Target Gene Details
Entrez Gene ID: | 2113 |
Gene Name: | ETS proto-oncogene 1, transcription factor |
Gene Aliases: |
ETS-1, EWSR2, c-ets-1, p54 |
Location: |
Chr.11:128458761-128587593 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETS1 | NM_001143820.1 | 8 | 1107 | NP_001137292.1 |
| NM_001162422.1 | 2 | 558 | NP_001155894.1 | |
| NM_005238.3 | 6 | 1206 | NP_005229.1 | |
| XM_011542649.2 | 9 | 1243 | XP_011540951.1 | |
| XM_011542650.1 | 7 | 1698 | XP_011540952.1 | |
| XM_011542651.1 | XP_011540953.1 | |||
| XM_011542652.1 | XP_011540954.1 | |||
| XM_017017314.1 | 8 | 1096 | XP_016872803.1 | |
| XM_017017315.1 | 7 | 1022 | XP_016872804.1 | |
| XM_017017316.1 | 7 | 17213 | XP_016872805.1 | |
| XM_017017317.1 | XP_016872806.1 | |||
| AK291840.1 | 6 | 1200 | ||
| AK301405.1 | ||||
| AY943926.1 | 2 | 242 | ||
| BC017314.2 | AAH17314.1 | |||
| BT019452.1 | 6 | 890 | AAV38259.1 | |
| BT019453.1 | 6 | 890 | AAV38260.1 | |
| BX640634.1 | 8 | 1107 | CAE45783.1 | |
| CR542254.1 | 6 | 890 | CAG47050.1 | |
| J04101.1 | 6 | 1009 | AAA52410.1 | |
| X14798.1 | 6 | 1168 | CAA32904.1 |
More Information
Set Membership: |
|

Genomic Map