Assay Details
Target Gene Details
Entrez Gene ID: | 4863 |
Gene Name: | nuclear protein, coactivator of histone transcription |
Gene Aliases: |
E14, E14/NPAT, p220 |
Location: |
Chr.11:108155280-108222638 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAT | NM_001321307.1 | 13 | 2056 | NP_001308236.1 |
| NM_002519.2 | 13 | 2056 | NP_002510.2 | |
| XM_011542854.2 | 13 | 2056 | XP_011541156.1 | |
| XM_017017802.1 | 13 | 2794 | XP_016873291.1 | |
| XM_017017803.1 | 13 | 2237 | XP_016873292.1 | |
| AK290020.1 | 13 | 2056 | ||
| AK291687.1 | 13 | 1980 | ||
| BC040356.1 | 13 | 2047 | AAH40356.1 | |
| BC050561.2 | 13 | 2041 | AAH50561.1 | |
| BC136270.1 | 13 | 1977 | ||
| D83243.1 | 13 | 2020 | BAA11861.1 | |
| U58852.1 | 13 | 1954 | AAB02735.1 | |
| X97186.1 | 13 | 1988 | CAA65824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1278n100 | Chr.11:108079845 - 108178945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map