Assay Details
Target Gene Details
Entrez Gene ID: | 490 |
Gene Name: | ATPase plasma membrane Ca2+ transporting 1 |
Gene Aliases: |
PMCA1, PMCA1kb |
Location: |
Chr.12:89588049-89709353 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2B1 | NM_001001323.1 | 21 | 6286 | NP_001001323.1 |
| NM_001682.2 | 20 | 6132 | NP_001673.2 | |
| XM_017019357.1 | 20 | 6063 | XP_016874846.1 | |
| XM_017019358.1 | 19 | 5976 | XP_016874847.1 | |
| XM_017019359.1 | 20 | 6130 | XP_016874848.1 | |
| AF070606.1 | 1 | 1183 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832479 | Chr.12:89467313 - 89672992 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map