Assay Details
Target Gene Details
Entrez Gene ID: | 54363 |
Gene Name: | hydroxyacid oxidase 1 |
Gene Aliases: |
GOX, GOX1, HAOX1 |
Location: |
Chr.20:7882984-7940446 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAO1 | NM_017545.2 | NP_060015.1 | ||
| AB024079.1 | BAA82872.1 | |||
| AF231916.1 | AAF40199.1 | |||
| AF244134.1 | AAF63219.1 | |||
| AK289423.1 | ||||
| AK308963.1 | 7 | 1097 | ||
| AK313839.1 | ||||
| AL121739.1 | CAB57329.1 | |||
| BC113665.1 | ||||
| BC113667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059059 | Chr.20:7793987 - 8053708 on Build GRCh38 | Gain |
|
| nsv585384 | Chr.20:7571617 - 8336371 on Build GRCh38 | Gain |
|
| esv3893279 | Chr.20:7582024 - 7999743 on Build GRCh38 | Gain |
|
| nsv833915 | Chr.20:7798839 - 7957303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map