Assay Details
Target Gene Details
| Entrez Gene ID: | 101928033 | 
| Gene Name: | uncharacterized LOC101928033 | 
| Gene Aliases: | - | 
| Location: | Chr.5:170389493-170422844 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CTD-2270F17.1 | NR_109899.1 | 1 | 53 | |
| AK311026.1 | 1 | 53 | 
Target Gene Details
| Entrez Gene ID: | 30820 | 
| Gene Name: | potassium voltage-gated channel interacting protein 1 | 
| Gene Aliases: | KCHIP1, VABP | 
| Location: | Chr.5:170353055-170736632 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KCNIP1 | NM_001034838.2 | NP_001030010.1 | ||
| XM_017009407.1 | XP_016864896.1 | |||
| XM_017009408.1 | XP_016864897.1 | |||
| AK301775.1 | ||||
| DC420716.1 | ||||
| DQ148476.1 | AAZ77793.1 | 
Target Gene Details
| Entrez Gene ID: | 3779 | 
| Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 1 | 
| Gene Aliases: | BKbeta1, K(VCA)beta, SLO-BETA, hbeta1, hslo-beta, k(VCA)beta-1, slo-beta-1 | 
| Location: | Chr.5:170378161-170389677 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KCNMB1 | NM_004137.3 | 1 | 133 | NP_004128.1 | 
| AK313979.1 | 1 | 111 | ||
| BC025707.1 | 1 | 90 | AAH25707.1 | |
| DA619309.1 | 1 | 133 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv471054 | Chr.5:170278287 - 170532433 on Build GRCh38 | Loss |  KCNIP1  LINC01366  CTD-2270F17.1  LCP2  KCNMB1 | 
| nsv600260 | Chr.5:170265550 - 170532433 on Build GRCh38 | Loss |  KCNIP1  LINC01366  CTD-2270F17.1  LCP2  KCNMB1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top