Assay Details
Target Gene Details
| Entrez Gene ID: | 83642 | 
| Gene Name: | selenoprotein O | 
| Gene Aliases: | - | 
| Location: | Chr.22:50200979-50217616 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| SELO | NM_031454.1 | NP_113642.1 | ||
| AY324823.1 | AAP85540.1 | |||
| BC001099.2 | AAH01099.3 | |||
| BC020510.1 | AAH20510.1 | |||
| BC110866.1 | AAI10867.1 | |||
| CR456464.1 | CAG30350.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv955199 | Chr.22:50149372 - 50320271 on Build GRCh38 | Deletion |  TRABD  MAPK12  PANX2  MAPK11  HDAC10  SELO  DENND6B  TUBGCP6  PLXNB2  MOV10L1 | 
| nsv471223 | Chr.22:50190317 - 50271919 on Build GRCh38 | Loss |  TRABD  MAPK12  MAPK11  HDAC10  SELO  TUBGCP6 | 
| dgv781n67 | Chr.22:50153213 - 50314433 on Build GRCh38 | Gain |  TRABD  MAPK12  PANX2  MAPK11  HDAC10  SELO  DENND6B  TUBGCP6  PLXNB2  MOV10L1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic