Assay Details
Target Gene Details
Entrez Gene ID: | 4193 |
Gene Name: | MDM2 proto-oncogene |
Gene Aliases: |
ACTFS, HDMX, hdm2 |
Location: |
Chr.12:68808149-68845544 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
More Information
Set Membership: |
|

Genomic Map