Assay Details
Target Gene Details
Entrez Gene ID: | 90 |
Gene Name: | activin A receptor type 1 |
Gene Aliases: |
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI |
Location: |
Chr.2:157736446-157875880 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACVR1 | NM_001105.4 | 11 | 2423 | NP_001096.1 |
| NM_001111067.2 | 11 | 2242 | NP_001104537.1 | |
| XM_005246939.3 | 11 | 2294 | XP_005246996.1 | |
| XM_005246940.3 | 11 | 2262 | XP_005246997.1 | |
| XM_006712825.3 | 11 | 2375 | XP_006712888.1 | |
| XM_011512106.2 | 12 | 2342 | XP_011510408.1 | |
| XM_011512107.2 | 11 | 2314 | XP_011510409.1 | |
| XM_011512108.2 | 11 | 3721 | XP_011510410.1 | |
| BC033867.1 | 11 | 2423 | AAH33867.1 | |
| Z22534.1 | 10 | 2096 | CAA80256.1 |
More Information
Set Membership: |
|

Genomic Map