Assay Details
Target Gene Details
Entrez Gene ID: | 120892 |
Gene Name: | leucine rich repeat kinase 2 |
Gene Aliases: |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
Location: |
Chr.12:40224895-40369285 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRK2 | NM_198578.3 | 10 | 1252 | NP_940980.3 |
| XM_005268629.3 | 10 | 1368 | XP_005268686.1 | |
| XM_011537877.2 | 10 | 1368 | XP_011536179.1 | |
| XM_011537881.2 | 10 | 1368 | XP_011536183.1 | |
| XM_011537882.2 | 10 | 1368 | XP_011536184.1 | |
| XM_017018786.1 | 10 | 1368 | XP_016874275.1 | |
| XM_017018789.1 | 10 | 1368 | XP_016874278.1 | |
| AK127729.1 | ||||
| AY792511.1 | 10 | 1131 | AAV63975.1 | |
| BC117180.1 | 10 | 1189 | ||
| BM285390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819273 | Chr.12:40251264 - 40252159 on Build GRCh38 | Gain |
|
| nsv1048258 | Chr.12:39970608 - 41209299 on Build GRCh38 | Gain |
|
| nsv1042967 | Chr.12:40235834 - 40316517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map