Assay Details
Target Gene Details
Entrez Gene ID: | 9039 |
Gene Name: | ubiquitin like modifier activating enzyme 3 |
Gene Aliases: |
NAE2, UBE1C, hUBA3 |
Location: |
Chr.3:69054730-69080373 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA3 | NM_003968.3 | 18 | 1627 | NP_003959.3 |
| NM_198195.1 | 17 | 1585 | NP_937838.1 | |
| XM_011534210.1 | 17 | 1541 | XP_011532512.1 | |
| XM_011534211.1 | 16 | 1494 | XP_011532513.1 | |
| AF046024.1 | 17 | 1628 | AAC27648.1 | |
| AK002159.1 | 17 | 1572 | ||
| AL117566.1 | 18 | 1623 | CAB55996.2 | |
| BC022853.1 | 18 | 1626 | AAH22853.1 | |
| CR533537.1 | CAG38568.1 |
More Information
Set Membership: |
|

Genomic Map